siRNA Details
| Gene Symbol: Mipol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164370.1 | NP_001157842.1 | 3 | 481 | ||
| GenBank mRNA | AK006974.1 | - | 390 | |||
| AK044766.1 | - | 2 | 208 | |||
| AK046422.1 | - | 3 | 479 | |||
| AK050030.1 | - | 3 | 467 | |||
| AK077915.1 | - | 3 | 461 | |||
| AK085510.1 | - | 3 | 476 | |||
| AK163490.1 | - | 3 | 461 | |||
| AK168989.1 | - | 2 | 266 | |||

Mipol1
Genomic Map