siRNA Details
| Gene Symbol: 1700091H14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007033.1 | - | 2 | 273 | ||
| Gene Symbol: Gm17026 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076616.1 | - | 2 | 250 | ||

1700091H14Rik
Genomic Map