siRNA Details
| Gene Symbol: Vgll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028572.1 | NP_082848.1 | 4 | 1297 | ||
| GenBank mRNA | AK007165.1 | - | 3 | 984 | ||
| BC042696.1 | - | 4 | 1222 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vgll3| Gene Symbol: Vgll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028572.1 | NP_082848.1 | 4 | 1297 | ||
| GenBank mRNA | AK007165.1 | - | 3 | 984 | ||
| BC042696.1 | - | 4 | 1222 | |||