siRNA Details
| Gene Symbol: Trp53tg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271575.1 | NP_001258504.1 | 5 | 900 | ||
| GenBank mRNA | AK007294.1 | - | 5 | 901 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trp53tg5| Gene Symbol: Trp53tg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271575.1 | NP_001258504.1 | 5 | 900 | ||
| GenBank mRNA | AK007294.1 | - | 5 | 901 | ||