siRNA Details
| Gene Symbol: Zfp433 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243067.1 | NP_001229996.1 | 4 | 1445 | ||
| GenBank mRNA | BC064812.1 | - | 4 | 1444 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp433| Gene Symbol: Zfp433 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243067.1 | NP_001229996.1 | 4 | 1445 | ||
| GenBank mRNA | BC064812.1 | - | 4 | 1444 | ||