siRNA Details
| Gene Symbol: 1700125H20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028589.1 | NP_082865.1 | 5, 6 | 710 | ||
| GenBank mRNA | AK007281.1 | - | 4, 5 | 432 | ||
| BC048674.1 | - | 5, 6 | 710 | 6774125 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700125H20Rik| Gene Symbol: 1700125H20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028589.1 | NP_082865.1 | 5, 6 | 710 | ||
| GenBank mRNA | AK007281.1 | - | 4, 5 | 432 | ||
| BC048674.1 | - | 5, 6 | 710 | 6774125 | ||