siRNA Details
| Gene Symbol: 2410004P03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201332.1 | NP_001188261.1 | 5 | 1296 | ||
| NM_001201333.1 | NP_001188262.1 | 4 | 1256 | |||
| GenBank mRNA | AK010406.1 | - | 5 | 1121 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2410004P03Rik| Gene Symbol: 2410004P03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201332.1 | NP_001188261.1 | 5 | 1296 | ||
| NM_001201333.1 | NP_001188262.1 | 4 | 1256 | |||
| GenBank mRNA | AK010406.1 | - | 5 | 1121 | ||