siRNA Details
| Gene Symbol: Rec114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028598.1 | NP_082874.1 | 6 | 772 | ||
| GenBank mRNA | AK010725.1 | - | 6 | 769 | ||
| AK019702.1 | - | 3 | 400 | |||
| BC038688.1 | - | 6 | 769 | |||
| BY715467.1 | - | 3 | 398 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rec114| Gene Symbol: Rec114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028598.1 | NP_082874.1 | 6 | 772 | ||
| GenBank mRNA | AK010725.1 | - | 6 | 769 | ||
| AK019702.1 | - | 3 | 400 | |||
| BC038688.1 | - | 6 | 769 | |||
| BY715467.1 | - | 3 | 398 | |||