siRNA Details
| Gene Symbol: Rec114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028598.1 | NP_082874.1 | 4 | 366 | ||
| GenBank mRNA | AK010725.1 | - | 4 | 363 | ||
| AV474073.1 | - | 4 | 366 | |||
| BC038688.1 | - | 4 | 363 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rec114| Gene Symbol: Rec114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028598.1 | NP_082874.1 | 4 | 366 | ||
| GenBank mRNA | AK010725.1 | - | 4 | 363 | ||
| AV474073.1 | - | 4 | 366 | |||
| BC038688.1 | - | 4 | 363 | |||