siRNA Details
| Gene Symbol: Psma8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163609.1 | NP_001157081.1 | 7 | 802 | ||
| GenBank mRNA | AK010717.1 | - | 7 | 803 | ||
| BC116990.1 | - | 7 | 764 | 40126309 | ||
| BC116992.1 | - | 7 | 764 | 40126311 | ||
| BC144771.1 | - | 8 | 835 | 9053319 | ||
| BC144772.1 | - | 6 | 644 | 9053320 | ||

Psma8
Genomic Map