siRNA Details
| Gene Symbol: Atg16l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111111.1 | NP_001104581.1 | 18 | 2087 | ||
| GenBank mRNA | AK010773.1 | - | 17 | 2008 | ||
| AK131109.1 | - | 2021 | ||||
| AK155125.1 | - | 18 | 2077 | |||
| AK157410.1 | - | 18 | 2067 | |||
| AK158186.1 | - | 17 | 2196 | |||
| BC060054.1 | - | 17 | 2583 | |||

Atg16l2
Genomic Map