siRNA Details
| Gene Symbol: Atg16l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111111.1 | NP_001104581.1 | 13 | 1331 | ||
| GenBank mRNA | AB476647.1 | - | 12 | 1207 | ||
| AB476648.1 | - | 13 | 1270 | |||
| AK010773.1 | - | 12 | 1252 | |||
| AK131109.1 | - | 1101 | ||||
| AK155125.1 | - | 13 | 1321 | |||
| AK157410.1 | - | 13 | 1310 | |||
| AK158186.1 | - | 12 | 1445 | |||
| BC060054.1 | - | 12 | 1668 | |||

Atg16l2
Genomic Map