siRNA Details
| Gene Symbol: 2410089E03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162906.1 | NP_001156378.1 | 1 | 350 | ||
| GenBank mRNA | AK028897.1 | - | 1 | 351 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2410089E03Rik| Gene Symbol: 2410089E03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162906.1 | NP_001156378.1 | 1 | 350 | ||
| GenBank mRNA | AK028897.1 | - | 1 | 351 | ||