siRNA Details
| Gene Symbol: Gucy2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081076.2 | NP_001074545.1 | 17 | 2663 | ||
| GenBank mRNA | AK010727.1 | - | 2 | 100 | ||
| AK049940.1 | - | 5 | 682 | |||
| AY395631.1 | - | 17 | 2663 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gucy2g| Gene Symbol: Gucy2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081076.2 | NP_001074545.1 | 17 | 2663 | ||
| GenBank mRNA | AK010727.1 | - | 2 | 100 | ||
| AK049940.1 | - | 5 | 682 | |||
| AY395631.1 | - | 17 | 2663 | |||