siRNA Details
| Gene Symbol: 1110017D15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048005.1 | NP_001041470.1 | 1 | 217 | ||
| NM_001253777.1 | NP_001240706.1 | 1 | 217 | |||
| NM_001253788.1 | NP_001240717.1 | 1 | 217 | |||
| GenBank mRNA | AB231294.1 | - | 1 | 216 | ||
| BU939732.1 | - | 220 | ||||

1110017D15Rik
Genomic Map