siRNA Details
| Gene Symbol: Zfp383 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243908.1 | NP_001230837.1 | 5 | 2561 | ||
| GenBank mRNA | AK003357.1 | - | 1 | 1193 | ||
| BC064456.1 | - | 4 | 3149 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp383| Gene Symbol: Zfp383 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243908.1 | NP_001230837.1 | 5 | 2561 | ||
| GenBank mRNA | AK003357.1 | - | 1 | 1193 | ||
| BC064456.1 | - | 4 | 3149 | |||