siRNA Details
| Gene Symbol: 1110034G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028637.1 | NP_082913.1 | 1 | 128 | ||
| GenBank mRNA | BC100475.1 | - | 1 | 128 | 30253755 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

1110034G24Rik| Gene Symbol: 1110034G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028637.1 | NP_082913.1 | 1 | 128 | ||
| GenBank mRNA | BC100475.1 | - | 1 | 128 | 30253755 | |