siRNA Details
| Gene Symbol: 1110034G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028637.1 | NP_082913.1 | 3 | 748 | ||
| GenBank mRNA | AK004090.1 | - | 3 | 322 | ||
| BC100475.1 | - | 3 | 748 | 30253755 | ||
| BC138171.1 | - | 3 | 402 | 8861190 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008762239, XM_008762240.

1110034G24Rik
Genomic Map