siRNA Details
| Gene Symbol: Tex36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028654.1 | NP_082930.1 | 4 | 623 | ||
| GenBank mRNA | AK015080.1 | - | 4 | 668 | ||
| BC100509.1 | - | 4 | 623 | 30918228 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tex36| Gene Symbol: Tex36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028654.1 | NP_082930.1 | 4 | 623 | ||
| GenBank mRNA | AK015080.1 | - | 4 | 668 | ||
| BC100509.1 | - | 4 | 623 | 30918228 | ||