siRNA Details
Gene Symbol: Satl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028655.1 | NP_082931.1 | 1 | 485 | ||
GenBank mRNA | BC138009.1 | - | 1 | 485 | 8861028 | |
BC138011.1 | - | 1 | 485 | 8861030 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Satl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028655.1 | NP_082931.1 | 1 | 485 | ||
GenBank mRNA | BC138009.1 | - | 1 | 485 | 8861028 | |
BC138011.1 | - | 1 | 485 | 8861030 |