siRNA Details
Gene Symbol: Ankrd45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028664.1 | NP_082940.1 | 7 | 991 | ||
GenBank mRNA | AK016757.1 | - | 6 | 910 | ||
BC049713.1 | - | 7 | 963 | 6741553 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ankrd45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028664.1 | NP_082940.1 | 7 | 991 | ||
GenBank mRNA | AK016757.1 | - | 6 | 910 | ||
BC049713.1 | - | 7 | 963 | 6741553 |