siRNA Details
Gene Symbol: 4930415F15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028669.1 | NP_082945.1 | 6 | 653 | ||
GenBank mRNA | AK015144.1 | - | 6 | 653 | ||
AV254343.1 | - | 2 | 136 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930415F15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028669.1 | NP_082945.1 | 6 | 653 | ||
GenBank mRNA | AK015144.1 | - | 6 | 653 | ||
AV254343.1 | - | 2 | 136 |