siRNA Details
| Gene Symbol: Stk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019206.2 | NP_062079.2 | 21 | 2958 | ||
| XM_008767561.1 | XP_008765783.1 | 18 | 2962 | |||
| GenBank mRNA | U33472.1 | - | 13 | 2110 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stk10| Gene Symbol: Stk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019206.2 | NP_062079.2 | 21 | 2958 | ||
| XM_008767561.1 | XP_008765783.1 | 18 | 2962 | |||
| GenBank mRNA | U33472.1 | - | 13 | 2110 | ||