siRNA Details
| Gene Symbol: Otud4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081164.1 | NP_001074633.1 | 21 | 5829 | ||
| NM_001256033.1 | NP_001242962.1 | 21 | 5832 | |||
| GenBank mRNA | AK034477.1 | - | 1 | 2205 | ||
| AK078157.1 | - | 1 | 2025 | |||
| AK084296.1 | - | 1 | 1586 | |||
| AK122429.1 | - | 16 | 5123 | |||
| AK169183.1 | - | 1 | 626 | |||
| BC042641.1 | - | 1 | 2570 | |||
| BC138373.1 | - | 21 | 5804 | 8861393 | ||
| BC145259.1 | - | 21 | 5801 | 9053812 | ||

Otud4
Genomic Map