siRNA Details
| Gene Symbol: Jakmip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028708.2 | NP_082984.1 | 4075 | |||
| GenBank mRNA | AK132869.1 | - | 2 | 725 | ||
| AK220482.1 | - | 4055 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jakmip3| Gene Symbol: Jakmip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028708.2 | NP_082984.1 | 4075 | |||
| GenBank mRNA | AK132869.1 | - | 2 | 725 | ||
| AK220482.1 | - | 4055 | ||||