siRNA Details
| Gene Symbol: Steap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103156.2 | NP_001096626.1 | 6 | 6734 | ||
| NM_001103157.2 | NP_001096627.1 | 6 | 7089 | |||
| NM_001285469.1 | NP_001272398.1 | 6 | 6643 | |||
| NM_001285470.1 | NP_001272399.1 | 5 | 6608 | |||
| NM_028734.5 | NP_083010.2 | 6 | 6791 | |||

Steap2
Genomic Map