siRNA Details
| Gene Symbol: Serpina3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167705.1 | NP_001161177.1 | 5 | 1564 | ||
| NM_028740.2 | NP_083016.1 | 4 | 904 | |||
| GenBank mRNA | AK016702.1 | - | 4 | 907 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serpina3a| Gene Symbol: Serpina3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167705.1 | NP_001161177.1 | 5 | 1564 | ||
| NM_028740.2 | NP_083016.1 | 4 | 904 | |||
| GenBank mRNA | AK016702.1 | - | 4 | 907 | ||