siRNA Details
| Gene Symbol: Men1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019208.3 | NP_062081.1 | 7 | 1478 | ||
| XM_006230769.2 | XP_006230831.1 | 8 | 1196 | |||
| XM_006230770.2 | XP_006230832.1 | 8 | 1287 | |||
| XM_006230771.2 | XP_006230833.1 | 8 | 1197 | |||
| XM_006230772.2 | XP_006230834.1 | 8 | 1255 | |||
| GenBank mRNA | AB023400.1 | - | 7 | 1545 | ||
| AF130369.1 | - | 7 | 1453 | |||
| AF130370.1 | - | 7 | 1098 | |||

Men1
Genomic Map