siRNA Details
Gene Symbol: Syce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143765.1 | NP_001137237.1 | 13 | 1363 | ||
GenBank mRNA | AK016694.1 | - | 13 | 1363 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Syce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143765.1 | NP_001137237.1 | 13 | 1363 | ||
GenBank mRNA | AK016694.1 | - | 13 | 1363 |