siRNA Details
| Gene Symbol: Syce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143765.1 | NP_001137237.1 | 13 | 1012 | ||
| GenBank mRNA | AK016694.1 | - | 13 | 1012 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syce1| Gene Symbol: Syce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143765.1 | NP_001137237.1 | 13 | 1012 | ||
| GenBank mRNA | AK016694.1 | - | 13 | 1012 | ||