siRNA Details
Gene Symbol: 1700017B05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028820.2 | NP_083096.1 | 3 | 4669 | ||
GenBank mRNA | AK029230.1 | - | 4673 | |||
BC118527.1 | - | 3 | 4523 | 40098810 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700017B05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028820.2 | NP_083096.1 | 3 | 4669 | ||
GenBank mRNA | AK029230.1 | - | 4673 | |||
BC118527.1 | - | 3 | 4523 | 40098810 |