siRNA Details
| Gene Symbol: 1700016H13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163550.1 | NP_001157022.1 | 4 | 506 | ||
| NM_028824.1 | NP_083100.1 | 4 | 657 | |||
| GenBank mRNA | AK006033.1 | - | 4 | 659 | ||
| BC049644.1 | - | 4 | 649 | 6705673 | ||

1700016H13Rik
Genomic Map