siRNA Details
| Gene Symbol: Gsdmcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006248.1 | - | 5 | 1117 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gsdmcl1| Gene Symbol: Gsdmcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006248.1 | - | 5 | 1117 | ||