siRNA Details
| Gene Symbol: Iqcf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146701.1 | NP_001140173.1 | 1, 2 | 73 | ||
| GenBank mRNA | BC049769.1 | - | 52 | 6774620 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iqcf1| Gene Symbol: Iqcf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146701.1 | NP_001140173.1 | 1, 2 | 73 | ||
| GenBank mRNA | BC049769.1 | - | 52 | 6774620 | ||