siRNA Details
| Gene Symbol: Iqcf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146701.1 | NP_001140173.1 | 2 | 383 | ||
| NM_028843.1 | NP_083119.1 | 3 | 488 | |||
| GenBank mRNA | AK006797.1 | - | 3 | 464 | ||
| BC049769.1 | - | 362 | 6774620 | |||

Iqcf1
Genomic Map