siRNA Details
Gene Symbol: 1700080O16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028851.1 | NP_083127.1 | 4 | 1374 | ||
GenBank mRNA | AK006959.1 | - | 4 | 1375 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700080O16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028851.1 | NP_083127.1 | 4 | 1374 | ||
GenBank mRNA | AK006959.1 | - | 4 | 1375 |