siRNA Details
| Gene Symbol: 1700080O16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028851.1 | NP_083127.1 | 4 | 978 | ||
| GenBank mRNA | AK006959.1 | - | 4 | 979 | ||
| BC109337.2 | - | 4 | 953 | 40052428 | ||
| BC109338.1 | - | 3 | 836 | 40052437 | ||

1700080O16Rik
Genomic Map