siRNA Details
| Gene Symbol: Spem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028855.1 | NP_083131.1 | 2 | 248 | ||
| GenBank mRNA | AK007080.1 | - | 2 | 264 | ||
| BC048760.1 | - | 2 | 248 | |||
| BC100505.1 | - | 2 | 240 | 30917608 | ||
| BC144838.1 | - | 1 | 441 | |||
| EF120626.1 | - | 2 | 248 | |||

Spem1
Genomic Map