siRNA Details
| Gene Symbol: 1700106J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028859.1 | NP_083135.1 | 1 | 843 | ||
| GenBank mRNA | AK137123.1 | - | 1 | 845 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700106J16Rik| Gene Symbol: 1700106J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028859.1 | NP_083135.1 | 1 | 843 | ||
| GenBank mRNA | AK137123.1 | - | 1 | 845 | ||