siRNA Details
| Gene Symbol: Aes | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019220.1 | NP_062093.1 | 3, 4 | 325 | ||
| GenBank mRNA | BC101855.1 | - | 290 | 7933368 | ||
| L14462.1 | - | 3, 4 | 325 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aes| Gene Symbol: Aes | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019220.1 | NP_062093.1 | 3, 4 | 325 | ||
| GenBank mRNA | BC101855.1 | - | 290 | 7933368 | ||
| L14462.1 | - | 3, 4 | 325 | |||