siRNA Details
| Gene Symbol: 4933406J08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028914.1 | NP_083190.1 | 7 | 1286 | ||
| GenBank mRNA | AK016695.1 | - | 7 | 1287 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933406J08Rik| Gene Symbol: 4933406J08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028914.1 | NP_083190.1 | 7 | 1286 | ||
| GenBank mRNA | AK016695.1 | - | 7 | 1287 | ||