siRNA Details
| Gene Symbol: Ttc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028918.2 | NP_083194.2 | 12 | 2326 | ||
| GenBank mRNA | AK161541.1 | - | 11 | 2180 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc25| Gene Symbol: Ttc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028918.2 | NP_083194.2 | 12 | 2326 | ||
| GenBank mRNA | AK161541.1 | - | 11 | 2180 | ||