siRNA Details
| Gene Symbol: 4933402D24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256158.1 | NP_001243087.1 | 3 | 1377 | ||
| GenBank mRNA | AK016612.1 | - | 3 | 1378 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933402D24Rik| Gene Symbol: 4933402D24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256158.1 | NP_001243087.1 | 3 | 1377 | ||
| GenBank mRNA | AK016612.1 | - | 3 | 1378 | ||