siRNA Details
| Gene Symbol: Cmip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163262.1 | NP_001156734.1 | 21 | 3918 | ||
| NM_028941.1 | NP_083217.1 | 21 | 3741 | |||
| GenBank mRNA | AK020023.1 | - | 1 | 314 | ||
| AK049761.1 | - | 1 | 1177 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cmip| Gene Symbol: Cmip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163262.1 | NP_001156734.1 | 21 | 3918 | ||
| NM_028941.1 | NP_083217.1 | 21 | 3741 | |||
| GenBank mRNA | AK020023.1 | - | 1 | 314 | ||
| AK049761.1 | - | 1 | 1177 | |||