siRNA Details
| Gene Symbol: Slc1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289941.1 | NP_001276870.1 | 9 | 1697 | ||
| NM_001289942.1 | NP_001276871.1 | 9 | 1689 | |||
| NM_001289943.1 | NP_001276872.1 | 8 | 1488 | |||
| NM_019225.2 | NP_062098.1 | 10 | 1814 | |||
| GenBank mRNA | AF265360.1 | - | 9 | 1536 | ||
| S59158.1 | - | 10 | 1756 | |||

Slc1a3
Genomic Map