siRNA Details
| Gene Symbol: 4933433C11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028961.1 | NP_083237.1 | 6 | 1076 | ||
| GenBank mRNA | AK017033.1 | - | 5 | 1166 | ||
| AK029738.1 | - | 6 | 1034 | |||
| BC050147.1 | - | 2 | 623 | |||
| BC132286.1 | - | 6 | 1076 | 40130563 | ||
| BC138006.1 | - | 6 | 1076 | 8861025 | ||

4933433C11Rik
Genomic Map