siRNA Details
Gene Symbol: Themis3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028998.1 | NP_083274.1 | 4 | 1048 | ||
GenBank mRNA | AK018663.1 | - | 4 | 1049 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Themis3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028998.1 | NP_083274.1 | 4 | 1048 | ||
GenBank mRNA | AK018663.1 | - | 4 | 1049 |