siRNA Details
| Gene Symbol: Nkx6-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029002.2 | NP_083278.1 | 3 | 1516 | ||
| GenBank mRNA | AK018683.1 | - | 3 | 1517 | ||
| AK147163.1 | - | 3 | 1488 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nkx6-3| Gene Symbol: Nkx6-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029002.2 | NP_083278.1 | 3 | 1516 | ||
| GenBank mRNA | AK018683.1 | - | 3 | 1517 | ||
| AK147163.1 | - | 3 | 1488 | |||