siRNA Details
| Gene Symbol: Itsn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136096.1 | NP_001129568.1 | 669 | |||
| NM_019227.1 | NP_062100.1 | 6 | 648 | |||
| GenBank mRNA | AF127798.1 | - | 6 | 648 | ||
| AF132672.1 | - | 6 | 648 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itsn1| Gene Symbol: Itsn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136096.1 | NP_001129568.1 | 669 | |||
| NM_019227.1 | NP_062100.1 | 6 | 648 | |||
| GenBank mRNA | AF127798.1 | - | 6 | 648 | ||
| AF132672.1 | - | 6 | 648 | |||