siRNA Details
| Gene Symbol: Slc12a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019229.2 | NP_062102.1 | 15 | 1944 | ||
| XM_006255482.2 | XP_006255544.1 | 15 | 3348 | |||
| XM_008772492.1 | XP_008770714.1 | 14 | 1935 | |||
| GenBank mRNA | U55815.1 | - | 15 | 1944 | ||

Slc12a4
Genomic Map